accessibility menu, dialog, popup

UserName

CAS RN 350-31-2

ONOClF

CAS RN 350-31-2

IUPAC Name: 1-chloro-2-fluoro-4-nitrobenzene
Synonyms: 1-chloro-2-fluoro-4-nitrobenzene
Molecular Weight (g/mol): 175.54
Molecular Formula: C6H3ClFNO2
InChi Key: CSSSAKOGRYYMSA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)C(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results