missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-fluoro-4-nitrobenzene 97.0+%, TCI America™

Catalog No. C21955G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21955G 5 g
C219525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21955G Supplier TCI America Supplier No. C21955G
Only null left

Chemical Identifiers

CAS 350-31-2
Molecular Formula C6H3ClFNO2
Molecular Weight (g/mol) 175.54
MDL Number MFCD00215833
InChI Key CSSSAKOGRYYMSA-UHFFFAOYSA-N
PubChem CID 67689
IUPAC Name 1-chloro-2-fluoro-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(Cl)C(F)=C1

Specifications

Melting Point 61°C
Boiling Point 116°C
Quantity 5 g
Formula Weight 175.54
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Chloro-2-fluoro-4-nitrobenzene