accessibility menu, dialog, popup

UserName

NaH3CH3CSOOOO

CAS RN 34850-66-3

IUPAC Name: sodium {7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl}methanesulfonate
Synonyms: sodium camphorsulfonate anion
Molecular Weight (g/mol): 254.28
Molecular Formula: C10H15NaO4S
InChi Key: AWMAOFAHBPCBHJ-UHFFFAOYNA-M
SMILES: [Na+].CC1(C)C2CCC1(CS([O-])(=O)=O)C(=O)C2
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)