CAS RN 3478-88-4
CAS RN 3478-88-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,2',4,4'-Tetramethylbenzophenone 96.0+%, TCI America™
CAS: 3478-88-4 Molecular Formula: C17H18O Molecular Weight (g/mol): 238.33 MDL Number: MFCD00060094 InChI Key: KFNQSAKXSGGDAA-UHFFFAOYSA-N PubChem CID: 12274022 IUPAC Name: bis(2,4-dimethylphenyl)methanone SMILES: CC1=CC(C)=C(C=C1)C(=O)C1=C(C)C=C(C)C=C1