missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2',4,4'-Tetramethylbenzophenone 96.0+%, TCI America™
$186.41 - $561.08
Chemical Identifiers
| CAS | 3478-88-4 |
|---|---|
| Molecular Formula | C17H18O |
| Molecular Weight (g/mol) | 238.33 |
| MDL Number | MFCD00060094 |
| InChI Key | KFNQSAKXSGGDAA-UHFFFAOYSA-N |
| PubChem CID | 12274022 |
| IUPAC Name | bis(2,4-dimethylphenyl)methanone |
| SMILES | CC1=CC(C)=C(C=C1)C(=O)C1=C(C)C=C(C)C=C1 |
Chemical Identifiers
| 3478-88-4 | |
| 238.33 | |
| KFNQSAKXSGGDAA-UHFFFAOYSA-N | |
| bis(2,4-dimethylphenyl)methanone |
| C17H18O | |
| MFCD00060094 | |
| 12274022 | |
| CC1=CC(C)=C(C=C1)C(=O)C1=C(C)C=C(C)C=C1 |
Specifications
| 3478-88-4 | |
| 188°C | |
| MFCD00060094 | |
| KFNQSAKXSGGDAA-UHFFFAOYSA-N | |
| bis(2,4-dimethylphenyl)methanone | |
| 12274022 | |
| ≥96.0% (GC) | |
| 2,2′,4,4′-Tetramethylbenzophenone |
| Yellow | |
| C17H18O | |
| 5 g | |
| CC1=CC(C)=C(C=C1)C(=O)C1=C(C)C=C(C)C=C1 | |
| 238.33 | |
| 238.33 | |
| Liquid |
Safety and Handling
TSCA : No