accessibility menu, dialog, popup

UserName

CAS RN 3475-21-6

CH3N

CAS RN 3475-21-6

IUPAC Name: 4-methyl-2-phenylpyridine
Synonyms: 4-methyl-2-phenylpyridine
Molecular Weight (g/mol): 169.23
Molecular Formula: C12H11N
InChi Key: WWMRJCUZPJJWBC-UHFFFAOYSA-N
SMILES: CC1=CC=NC(=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results