accessibility menu, dialog, popup

UserName

CH3CH3MgBr

CAS RN 34696-73-6

IUPAC Name: bromo(3,5-dimethylphenyl)magnesium
Synonyms: bromo(3,5-dimethylphenyl)magnesium
Molecular Weight (g/mol): 209.37
Molecular Formula: C8H9BrMg
InChi Key: XPXQQJMQESWIKM-UHFFFAOYSA-M
SMILES: CC1=CC([Mg]Br)=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results