accessibility menu, dialog, popup

UserName

CAS RN 34589-46-3

IUPAC Name: bromo(2,4-dimethylphenyl)magnesium
Synonyms: bromo(2,4-dimethylphenyl)magnesium
Molecular Weight (g/mol): 209.37
Molecular Formula: C8H9BrMg
InChi Key: HPGWKALAJLXSDH-UHFFFAOYSA-M
SMILES: CC1=CC=C([Mg]Br)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results