missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals PARP Inhibitor VIII, PJ34

Catalog No. AAJ64413MB
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64413MB 25 mg
AAJ64413LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64413MB Supplier Thermo Scientific Chemicals Supplier No. J64413MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 344458-15-7 | C17H17N3O2·HCl, C17H18ClN3O2 | 331.80 g/mol

A cell-permeable, water-soluble phenanthridinone-derivative that acts as a potent inhibitor of poly(ADP-ribose) polymerase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A cell-permeable, water-soluble phenanthridinone-derivative that acts as a potent inhibitor of poly(ADP-ribose) polymerase

Chemical Identifiers

CAS 344458-15-7
Molecular Formula C17H17N3O2·HCl
Synonym 2-Dimethylamino-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide hydrochloride

Specifications

Quantity 25 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) A cell-permeable, water-soluble phenanthridinone-derivative that acts as a potent inhibitor of poly(ADP-ribose) polymerase
Formula Weight 331.80g/mol
Sensitivity Light Sensitive and Hygroscopic
Product Type Signal Transduction Reagent
Form Solid

RUO – Research Use Only