accessibility menu, dialog, popup

UserName

CAS RN 3437-95-4

IUPAC Name: 2-iodothiophene
Synonyms: 2-iodothiophene
Molecular Weight (g/mol): 210.03
Molecular Formula: C4H3IS
InChi Key: ROIMNSWDOJCBFR-UHFFFAOYSA-N
SMILES: IC1=CC=CS1
Boiling Point 
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)