accessibility menu, dialog, popup

UserName

CAS RN 34171-69-2

IUPAC Name: potassium tricyanomethanide
Synonyms: potassium tricyanomethanide
Molecular Weight (g/mol): 129.16
Molecular Formula: C4KN3
InChi Key: ZKJPYQKGNUBNOA-UHFFFAOYSA-N
SMILES: [K+].N#C[C-](C#N)C#N
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)