CAS RN 34125-84-3
IUPAC Name:
1-[(chloromethyl)sulfanyl]-4-methylbenzene
Synonyms:
1-[(chloromethyl)sulfanyl]-4-methylbenzene
Molecular Weight (g/mol):
172.67
Molecular Formula:
C8H9ClS
InChi Key:
VFQYMJYUSRAJCZ-UHFFFAOYSA-N
SMILES:
CC1=CC=C(SCCl)C=C1
1
–
1
of
1
results
1
Chloromethyl p-Tolyl Sulfide 96.0+%, TCI America™
CAS: 34125-84-3 Molecular Formula: C8H9ClS Molecular Weight (g/mol): 172.67 MDL Number: MFCD01318451 InChI Key: VFQYMJYUSRAJCZ-UHFFFAOYSA-N Synonym: 4-(Chloromethylthio)toluene, p-Tolyl Chloromethyl Sulfide PubChem CID: 1502059 IUPAC Name: 1-[(chloromethyl)sulfanyl]-4-methylbenzene SMILES: CC1=CC=C(SCCl)C=C1