CAS RN 34035-03-5
IUPAC Name:
5-(4-chlorophenyl)furan-2-carbaldehyde
Synonyms:
5-(4-chlorophenyl)furan-2-carbaldehyde
Molecular Weight (g/mol):
206.63
Molecular Formula:
C11H7ClO2
InChi Key:
ROJGJNINTRCMBL-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1)C1=CC=C(O1)C=O
1
–
1
of
1
results
1
Promotions Available
5-(4-Chlorophenyl)furfural, 95+%, Thermo Scientific™
5-(4-Chlorophenyl)furfural, 95+%, Thermo Scientific™
CAS: 34035-03-5 Molecular Formula: C11H7ClO2 Molecular Weight (g/mol): 206.63 MDL Number: MFCD00195947 InChI Key: ROJGJNINTRCMBL-UHFFFAOYSA-N Synonym: 5-4-chlorophenyl furan-2-carbaldehyde,5-4-chlorophenyl-2-furaldehyde,5-4-chlorophenyl furfural,5-4-chloro-phenyl-furan-2-carbaldehyde,2-formyl-5-4-chlorophenyl furan,2-furancarboxaldehyde, 5-4-chlorophenyl,5-4-chlorophenyl-2-furancarboxaldehyde,5-4-chlorophenyl furan-2-carboxaldehyde,pubchem4001,cambridge id 7179578 PubChem CID: 676184 IUPAC Name: 5-(4-chlorophenyl)furan-2-carbaldehyde SMILES: ClC1=CC=C(C=C1)C1=CC=C(O1)C=O