accessibility menu, dialog, popup

UserName

CAS RN 3403-25-6

IUPAC Name: tert-butyl 2-amino-3-phenylpropanoate hydrochloride
Synonyms: tert-butyl 2-amino-3-phenylpropanoate hydrochloride
Molecular Weight (g/mol): 257.76
Molecular Formula: C13H20ClNO2
InChi Key: FDMCEXDXULPJPG-UHFFFAOYNA-N
SMILES: Cl.CC(C)(C)OC(=O)C(N)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)