accessibility menu, dialog, popup

UserName

CAS RN 33904-03-9

IUPAC Name: 1-isothiocyanato-2,4-dimethoxybenzene
Synonyms: 1-isothiocyanato-2,4-dimethoxybenzene
Molecular Weight (g/mol): 195.24
Molecular Formula: C9H9NO2S
InChi Key: CNXSCEPTSXPBTP-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C=C1)N=C=S
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)