missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxyphenyl Isothiocyanate 98.0+%, TCI America™

Catalog No. D49435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49435G 5 g
D494325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49435G Supplier TCI America Supplier No. D49435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33904-03-9
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.236
MDL Number MFCD00041058
InChI Key CNXSCEPTSXPBTP-UHFFFAOYSA-N
Synonym 2,4-dimethoxyphenyl isothiocyanate, 1-isothiocyanato-2,4-dimethoxy-benzene, 2,4-dimethoxybenzenisothiocyanate, acmc-20aock, 2,4-dimethoxyphenylisothiocyanate, 1-isothiocyanato-2.4-dimethoxybenzene, isothiocyanic acid 2,4-dimethoxyphenyl ester
PubChem CID 2736202
IUPAC Name 1-isothiocyanato-2,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)N=C=S)OC

Specifications

Melting Point 54°C
Color Yellow
Boiling Point 167°C
Quantity 5 g
UN Number 1759
Formula Weight 195.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethoxyphenyl Isothiocyanate