accessibility menu, dialog, popup

UserName

CAS RN 33893-85-5

IUPAC Name: 4-(hydroxymethyl)cyclohexan-1-ol
Synonyms: 4-(hydroxymethyl)cyclohexan-1-ol
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: VGRZISGVNOKTQU-UHFFFAOYSA-N
SMILES: OCC1CCC(O)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results