CAS RN 3385-94-2
Bis(trimethylsilyl)sulfide, 98%
CAS: 3385-94-2 Molecular Formula: C6H18SSi2 Molecular Weight (g/mol): 178.44 MDL Number: MFCD00014851 InChI Key: RLECCBFNWDXKPK-UHFFFAOYSA-N Synonym: bis trimethylsilyl sulfide,hexamethyldisilthiane,hexamethyldisilathiane,disilathiane, hexamethyl,1,1,1,3,3,3-hexamethyldisilathiane,disilthiane, hexamethyl,disilathiane, 1,1,1,3,3,3-hexamethyl,thiobis trimethylsilane,acmc-1ckj8 PubChem CID: 76920 IUPAC Name: trimethyl(trimethylsilylsulfanyl)silane SMILES: C[Si](C)(C)S[Si](C)(C)C
Bis(trimethylsilyl)sulfide, tech.
CAS: 3385-94-2 Molecular Formula: C6H18SSi2 Molecular Weight (g/mol): 178.44 MDL Number: MFCD00014851 InChI Key: RLECCBFNWDXKPK-UHFFFAOYSA-N Synonym: bis trimethylsilyl sulfide,hexamethyldisilthiane,hexamethyldisilathiane,disilathiane, hexamethyl,1,1,1,3,3,3-hexamethyldisilathiane,disilthiane, hexamethyl,disilathiane, 1,1,1,3,3,3-hexamethyl,thiobis trimethylsilane,acmc-1ckj8 PubChem CID: 76920 IUPAC Name: trimethyl(trimethylsilylsulfanyl)silane SMILES: C[Si](C)(C)S[Si](C)(C)C
Bis(trimethylsilyl)sulfide, 95%
CAS: 3385-94-2 Molecular Formula: C6H18SSi2 Molecular Weight (g/mol): 178.45 MDL Number: MFCD00014851 InChI Key: RLECCBFNWDXKPK-UHFFFAOYSA-N Synonym: bis trimethylsilyl sulfide,hexamethyldisilthiane,hexamethyldisilathiane,disilathiane, hexamethyl,1,1,1,3,3,3-hexamethyldisilathiane,disilthiane, hexamethyl,disilathiane, 1,1,1,3,3,3-hexamethyl,thiobis trimethylsilane,acmc-1ckj8 PubChem CID: 76920 IUPAC Name: trimethyl(trimethylsilylsulfanyl)silane SMILES: C[Si](C)(C)S[Si](C)(C)C
Bis(trimethylsilyl) Sulfide 97.0+%, TCI America™
CAS: 3385-94-2 Molecular Formula: C6H18SSi2 Molecular Weight (g/mol): 178.44 MDL Number: MFCD00014851 InChI Key: RLECCBFNWDXKPK-UHFFFAOYSA-N Synonym: bis trimethylsilyl sulfide,hexamethyldisilthiane,hexamethyldisilathiane,disilathiane, hexamethyl,1,1,1,3,3,3-hexamethyldisilathiane,disilthiane, hexamethyl,disilathiane, 1,1,1,3,3,3-hexamethyl,thiobis trimethylsilane,acmc-1ckj8 PubChem CID: 76920 IUPAC Name: trimethyl(trimethylsilylsulfanyl)silane SMILES: C[Si](C)(C)S[Si](C)(C)C