accessibility menu, dialog, popup

UserName

CAS RN 3385-21-5

IUPAC Name: (1R,3S)-cyclohexane-1,3-bis(aminium)
Synonyms: (1R,3S)-cyclohexane-1,3-bis(aminium)
Molecular Weight (g/mol): 116.21
Molecular Formula: C6H16N2
InChi Key: GEQHKFFSPGPGLN-OLQVQODUSA-P
SMILES: [NH3+][C@H]1CCC[C@@H]([NH3+])C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)