accessibility menu, dialog, popup

UserName

CAS RN 337508-54-0

SOONBr

CAS RN 337508-54-0

IUPAC Name: 1-(benzenesulfonyl)-2-(bromomethyl)-1H-indole
Synonyms: 1-(benzenesulfonyl)-2-(bromomethyl)indole
Molecular Weight (g/mol): 350.23
Molecular Formula: C15H12BrNO2S
InChi Key: SYYPBRKVYRCLJS-UHFFFAOYSA-N
SMILES: BrCC1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)