CAS RN 3364-80-5
IUPAC Name:
1,3-thiazole-4-carbaldehyde
Synonyms:
1,3-thiazole-4-carbaldehyde
Molecular Weight (g/mol):
113.13
Molecular Formula:
C4H3NOS
InChi Key:
WRFKSVINLIQRKF-UHFFFAOYSA-N
SMILES:
O=CC1=CSC=N1
1
–
1
of
1
results
1
Promotions Available
Thiazole-4-carboxaldehyde, 95%
Thiazole-4-carboxaldehyde, 95%
CAS: 3364-80-5 Molecular Formula: C4H3NOS Molecular Weight (g/mol): 113.13 MDL Number: MFCD00626896 InChI Key: WRFKSVINLIQRKF-UHFFFAOYSA-N Synonym: thiazole-4-carboxaldehyde,thiazole-4-carbaldehyde,4-thiazolecarboxaldehyde,thiazole-4-carboxyaldehyde,4-formylthiazole,1,3-thiazole-4-carboxaldehyde,4-thiazolecarbaldehyde,zlchem 1299,pubchem13812,4-formyl-1,3-thiazole PubChem CID: 2763214 IUPAC Name: 1,3-thiazole-4-carbaldehyde SMILES: O=CC1=CSC=N1