accessibility menu, dialog, popup

UserName

CH3OONONH3CH3CNH3CCH3NFPFFFFF(E/Z)

CAS RN 333717-40-1

IUPAC Name: ethyl cyano({[(dimethylamino)(dimethyliminiumyl)methoxy]imino})formate; hexafluoro-λ⁵-phosphanuide
Synonyms: ethyl cyano({[(dimethylamino)(dimethyliminio)methoxy]imino})formate hexafluorophosphate
Molecular Weight (g/mol): 386.24
Molecular Formula: C10H17F6N4O3P
InChi Key: RKTBAMPZUATMIO-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CCOC(=O)C(=NOC(N(C)C)=[N+](C)C)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results