accessibility menu, dialog, popup

UserName

CAS RN 33342-05-1

IUPAC Name: 1-cyclohexyl-3-{4-[2-(7-methoxy-4,4-dimethyl-1,3-dioxo-1,2,3,4-tetrahydroisoquinolin-2-yl)ethyl]benzenesulfonyl}urea
Synonyms: gliquidone glurenorm 格列喹酮
Molecular Weight (g/mol): 527.64
Molecular Formula: C27H33N3O6S
InChi Key: LLJFMFZYVVLQKT-UHFFFAOYSA-N
SMILES: COC1=CC=C2C(=C1)C(=O)N(CCC1=CC=C(C=C1)S(=O)(=O)NC(=O)NC1CCCCC1)C(=O)C2(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results