accessibility menu, dialog, popup

UserName

NH2NHNN

CAS RN 3325-11-9

IUPAC Name: 2H-1,2,3-benzotriazol-5-amine
Synonyms: 2H-1,2,3-benzotriazol-5-amine
Molecular Weight (g/mol): 134.14
Molecular Formula: C6H6N4
InChi Key: XSFHICWNEBCMNN-UHFFFAOYSA-N
SMILES: NC1=CC2=NNN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results