accessibility menu, dialog, popup

UserName

CAS RN 3321-03-7

IUPAC Name: 2-(2-aminoacetamido)-3-phenylpropanoic acid
Synonyms: 2-(2-aminoacetamido)-3-phenylpropanoic acid
Molecular Weight (g/mol): 222.24
Molecular Formula: C11H14N2O3
InChi Key: JBCLFWXMTIKCCB-UHFFFAOYNA-N
SMILES: NCC(=O)NC(CC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results