accessibility menu, dialog, popup

UserName

FFFIFFF

CAS RN 328-73-4

IUPAC Name: 1-iodo-3,5-bis(trifluoromethyl)benzene
Synonyms: 1-iodo-3,5-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 340.01
Molecular Formula: C8H3F6I
InChi Key: VDPIZIZDKPFXLI-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(I)=C1)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)