missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-3,5-bis(trifluoromethyl)benzene 98.0+%, TCI America™

Catalog No. B18685G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18685G 5 g
B186825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18685G Supplier TCI America Supplier No. B18685G

Chemical Identifiers

CAS 328-73-4
Molecular Formula C8H3F6I
Molecular Weight (g/mol) 340.007
MDL Number MFCD00040837
InChI Key VDPIZIZDKPFXLI-UHFFFAOYSA-N
Synonym 1-iodo-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl iodobenzene, 1,3-bis trifluoromethyl-5-iodobenzene, 3,5-bis trifluoromethyl-1-iodo benzene, benzene, 1-iodo-3,5-bis trifluoromethyl, 1-iodo-3,5-di trifluoromethyl benzene, 3-5-di trifluoromethyl iodobenzene, pubchem1056, acmc-209hwp
PubChem CID 630970
IUPAC Name 1-iodo-3,5-bis(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1C(F)(F)F)I)C(F)(F)F

Specifications

Color Yellow
Boiling Point 172°C
Quantity 5 g
Formula Weight 340.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodo-3,5-bis(trifluoromethyl)benzene