missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-3,5-bis(trifluoromethyl)benzene, 97+%

Catalog No. AAA1087814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1087814 25 g
AAA1087806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1087814 Supplier Thermo Scientific Chemicals Supplier No. A1087814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 328-73-4 | C8H3F6I | 340.007 g/mol

3,5-bis(trifluoromethyl)pheny1- magnesium iodide was prepared from 1-iodo-3,5- bis(trifluoromethyl)benzene. Monodeprotected masked hexayne was not particularly stable and hence was used directly without purification in a Sonogashira coupling with 1-iodo-3,5-bis(trifluoromethyl)benzene to provide mono-aryl-end-capped masked hexayne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-bis(trifluoromethyl)pheny1- magnesium iodide was prepared from 1-iodo-3,5- bis(trifluoromethyl)benzene. Monodeprotected masked hexayne was not particularly stable and hence was used directly without purification in a Sonogashira coupling with 1-iodo-3,5-bis(trifluoromethyl)benzene to provide mono-aryl-end-capped masked hexayne.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.

Chemical Identifiers

CAS 328-73-4
Molecular Formula C8H3F6I
Molecular Weight (g/mol) 340.007
MDL Number MFCD00040837
InChI Key VDPIZIZDKPFXLI-UHFFFAOYSA-N
Synonym 1-iodo-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl iodobenzene, 1,3-bis trifluoromethyl-5-iodobenzene, 3,5-bis trifluoromethyl-1-iodo benzene, benzene, 1-iodo-3,5-bis trifluoromethyl, 1-iodo-3,5-di trifluoromethyl benzene, 3-5-di trifluoromethyl iodobenzene, pubchem1056, acmc-209hwp
PubChem CID 630970
IUPAC Name 1-iodo-3,5-bis(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1C(F)(F)F)I)C(F)(F)F

Specifications

Density 1.919
Boiling Point 62°C to 64°C (12 mmHg)
Flash Point 74°C (165°F)
Refractive Index 1.4645
Quantity 25 g
Beilstein 2461627
Solubility Information Slightly soluble in water.
Formula Weight 340.01
Percent Purity ≥97%
Chemical Name or Material 1-Iodo-3,5-bis (trifluoromethyl)benzene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only