CAS RN 3279-90-1
IUPAC Name:
6-bromo-1,2,3,4-tetrahydroquinolin-2-one
Synonyms:
6-bromo-3,4-dihydro-1H-quinolin-2-one
Molecular Weight (g/mol):
226.07
Molecular Formula:
C9H8BrNO
InChi Key:
MQWZSSIUHXNNTM-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(NC(=O)CC2)C=C1
1
–
1
of
1
results
1
Promotions Available
6-Bromo-3,4-dihydro-2(1H)-quinolinone, 98%
6-Bromo-3,4-dihydro-2(1H)-quinolinone, 98%
CAS: 3279-90-1 Molecular Formula: C9H8BrNO Molecular Weight (g/mol): 226.07 MDL Number: MFCD03839832 InChI Key: MQWZSSIUHXNNTM-UHFFFAOYSA-N Synonym: 6-bromo-1,2,3,4-tetrahydro-2-quinolinone,6-bromo-3,4-dihydroquinolin-2 1h-one,6-bromo-3,4-dihydro-2 1h-quinolinone,6-bromo-3,4-dihydro-1h quinolin-2-one,6-bromo-1,2,3,4-tetrahydroquinolin-2-one,2 1h-quinolinone, 6-bromo-3,4-dihydro,6-bromo-2-oxo-1,2,3,4-tetrahydroquinoline,acmc-1co6x,6-bromo-3,4-dihydrocarbostyril,6-bromo-1,2,3,4-tetrahydroquinol-2-one PubChem CID: 14373281 IUPAC Name: 6-bromo-3,4-dihydro-1H-quinolin-2-one SMILES: BrC1=CC2=C(NC(=O)CC2)C=C1