accessibility menu, dialog, popup

UserName

CH3NH2

CAS RN 3261-62-9

IUPAC Name: 2-(4-methylphenyl)ethan-1-amine
Synonyms: 4-methylphenethylamine
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: VKJXAQYPOTYDLO-UHFFFAOYSA-N
SMILES: CC1=CC=C(CCN)C=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)