CAS RN 32462-30-9
CAS RN 32462-30-9
IUPAC Name:
(2S)-2-amino-2-(4-hydroxyphenyl)acetic acid
Synonyms:
oxfenicine L-4-hydroxyphenylglycine L-2-(4-hydroxyphenyl)glycin oxfenicina oxfenicinum
Molecular Weight (g/mol):
167.16
Molecular Formula:
C8H9NO3
InChi Key:
LJCWONGJFPCTTL-ZETCQYMHSA-N
SMILES:
N[C@H](C(O)=O)C1=CC=C(O)C=C1
1
–
1
of
1
results
1
4-Hydroxy-L-(+)-2-phenylglycine 99.0+%, TCI America™
CAS: 32462-30-9 Molecular Formula: C8H9NO3 Molecular Weight (g/mol): 167.164 MDL Number: MFCD00065932 InChI Key: LJCWONGJFPCTTL-ZETCQYMHSA-N Synonym: (S)-(+)-2-Amino-2-(4-hydroxyphenyl)acetic Acid, L-(+)-2-(4-Hydroxyphenyl)glycine, Oxfenicine, H-Phg(4-OH)-OH PubChem CID: 36143 ChEBI: CHEBI:31755 IUPAC Name: (2S)-2-amino-2-(4-hydroxyphenyl)acetic acid SMILES: C1=CC(=CC=C1C(C(=O)O)N)O