accessibility menu, dialog, popup

UserName

CAS RN 32372-82-0

IUPAC Name: 2,3-dihydro-1H-isoindole hydrochloride
Synonyms: isoindoline hydrochloride
Molecular Weight (g/mol): 155.63
Molecular Formula: C8H10ClN
InChi Key: NOVIRODZMIZUPA-UHFFFAOYSA-N
SMILES: Cl.C1NCC2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)