accessibility menu, dialog, popup

UserName

CAS RN 32315-10-9

IUPAC Name: ditrichloromethyl carbonate
Synonyms: triphosgene
Molecular Weight (g/mol): 296.73
Molecular Formula: C3Cl6O3
InChi Key: UCPYLLCMEDAXFR-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)OC(=O)OC(Cl)(Cl)Cl
Boiling Point 
  • (3)
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (3)
  • (4)
  • (2)
  • (5)
  • (4)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)