accessibility menu, dialog, popup

UserName

H3CCH3CH3

CAS RN 31807-55-3

IUPAC Name: 2-methylundecane
Synonyms: 2-methylundecane
Molecular Weight (g/mol): 170.34
Molecular Formula: C12H26
InChi Key: GTJOHISYCKPIMT-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(C)C
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)