accessibility menu, dialog, popup

UserName

OH(S)(R)(S)OO(R)OO

CAS RN 31752-99-5

IUPAC Name: (3aR,4S,5R,6aS)-4-(hydroxymethyl)-2-oxo-hexahydro-2H-cyclopenta[b]furan-5-yl [1,1'-biphenyl]-4-carboxylate
Synonyms: corey lactone 4-phenylbenzoate
Molecular Weight (g/mol): 352.39
Molecular Formula: C21H20O5
InChi Key: SZJVIFMPKWMGSX-AKHDSKFASA-N
SMILES: OC[C@H]1[C@@H](C[C@@H]2OC(=O)C[C@H]12)OC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results