accessibility menu, dialog, popup

UserName

CAS RN 31704-80-0

IUPAC Name: 2-(5-methylfuran-2-yl)butanal
Synonyms: 2-(5-methylfuran-2-yl)butanal
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: PFLUFBMKOXFWSD-UHFFFAOYNA-N
SMILES: CCC(C=O)C1=CC=C(C)O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results