accessibility menu, dialog, popup

UserName

CAS RN 317-34-0

IUPAC Name: bis(1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione); ethane-1,2-diamine
Synonyms: aminophylline phyllindon ammophyllin variaphylline theophyldine eurphyllin theophyllin aethylendiamin dura-tab S.M. aminophylline Dobo theolone
Molecular Weight (g/mol): 420.43
Molecular Formula: C16H24N10O4
InChi Key: FQPFAHBPWDRTLU-UHFFFAOYSA-N
SMILES: NCCN.CN1C2=C(NC=N2)C(=O)N(C)C1=O.CN1C2=C(NC=N2)C(=O)N(C)C1=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)