accessibility menu, dialog, popup

UserName

CAS RN 31643-49-9

IUPAC Name: 4-nitrobenzene-1,2-dicarbonitrile
Synonyms: 4-nitrobenzene-1,2-dicarbonitrile
Molecular Weight (g/mol): 173.13
Molecular Formula: C8H3N3O2
InChi Key: NTZMSBAAHBICLE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C#N)C(=C1)C#N
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)