missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophthalonitrile, 97%

Catalog No. AAH3354609
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3354609 10 g
AAH3354603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3354609 Supplier Thermo Scientific Chemicals Supplier No. H3354609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 31643-49-9 | C8H3N3O2 | 173.13 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 31643-49-9
Molecular Formula C8H3N3O2
Molecular Weight (g/mol) 173.13
MDL Number MFCD00040301
InChI Key NTZMSBAAHBICLE-UHFFFAOYSA-N
Synonym 4-nitrophthalonitrile, 5-nitrobenzene-1,2-dicarbonitrile, 4-nitro-1,2-benzenedicarbonitrile, 1,2-dicyano-4-nitrobenzene, 3,4-dicyanonitrobenzene, phthalonitrile, 4-nitro, 4-nitro-phthalonitrile, 1,2-benzenedicarbonitrile, 4-nitro, 5-nitro-1,2-benzenedicarbonitrile, 4-nitro-o-benzenedinitrile
PubChem CID 97443
IUPAC Name 4-nitrobenzene-1,2-dicarbonitrile
SMILES [O-][N+](=O)C1=CC=C(C#N)C(=C1)C#N

Specifications

Melting Point 136°C to 137°C
Quantity 10 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 173.13
Percent Purity 97%
Chemical Name or Material 4-Nitrophthalonitrile
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 250-748-9
RTECSNumber TI8576000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only