accessibility menu, dialog, popup

UserName

HO(R)O(R)(R)HO(S)HONFOHNO

CAS RN 316-46-1

IUPAC Name: 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5-fluorouridine 5-fluorouridine, 18F-labeled 5-fur FUR Fur FURD 18F-labeled 5-fluorouridine 5-fluorouridine, 18f-labeled 18f-labeled 5-fluorouridine F5130_sigma
Molecular Weight (g/mol): 262.19
Molecular Formula: C9H11FN2O6
InChi Key: FHIDNBAQOFJWCA-UAKXSSHOSA-N
SMILES: OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=C(F)C(=O)NC1=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)