missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorouridine, 97%

Catalog No. AAJ6208306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6208306 5 g
AAJ6208303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6208306 Supplier Thermo Scientific Chemicals Supplier No. J6208306

CAS: 316-46-1 | C9H11FN2O6 | 262.193 g/mol

5-Fluorouridine exhibits anticancer activity. It is often used in chemical and biochemical comparison studies with fluorouracil and thymine analogs. It is an active metabolite of Doxifluridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluorouridine exhibits anticancer activity. It is often used in chemical and biochemical comparison studies with fluorouracil and thymine analogs. It is an active metabolite of Doxifluridine.

Solubility
Soluble in Methanol (Slightly), Water (Slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Store at 4°C.

Chemical Identifiers

CAS 316-46-1
Molecular Formula C9H11FN2O6
Molecular Weight (g/mol) 262.193
MDL Number MFCD00036832
InChI Key FHIDNBAQOFJWCA-UAKXSSHOSA-N
Synonym 5-fluorouridine, 5-fur, uridine, 5-fluoro, furd, 5-fluoro-uridine, 5-fluorouracil 1beta-d-ribofuranoside, fluorouridine, chembl54918, 5-fluoro-1-beta-d-ribofuranosyl uracil, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-fluoropyrimidine-2,4 1h,3h-dione
PubChem CID 9427
ChEBI CHEBI:40154
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-fluoropyrimidine-2,4-dione
SMILES C1=C(C(=O)NC(=O)N1C2C(C(C(O2)CO)O)O)F

Specifications

Color White
Quantity 5 g
Beilstein 33662
Solubility Information Soluble in Methanol (Slightly),Water (Slightly).
Formula Weight 262.19
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 5-Fluorouridine
Hazard Category Warning
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 206-260-3
RTECSNumber YU8050000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only