missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorouridine 98.0+%, TCI America™

Catalog No. F06365G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06365G 5 g
F06361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06365G Supplier TCI America Supplier No. F06365G

Chemical Identifiers

CAS 316-46-1
Molecular Formula C9H11FN2O6
Molecular Weight (g/mol) 262.193
MDL Number MFCD00036832
InChI Key FHIDNBAQOFJWCA-UAKXSSHOSA-N
Synonym 5-fluorouridine, 5-fur, uridine, 5-fluoro, furd, 5-fluoro-uridine, 5-fluorouracil 1beta-d-ribofuranoside, fluorouridine, chembl54918, 5-fluoro-1-beta-d-ribofuranosyl uracil, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-fluoropyrimidine-2,4 1h,3h-dione
PubChem CID 9427
ChEBI CHEBI:40154
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-fluoropyrimidine-2,4-dione
SMILES C1=C(C(=O)NC(=O)N1C2C(C(C(O2)CO)O)O)F

Specifications

Melting Point 185°C
Color White-Yellow
Quantity 5 g
Formula Weight 262.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluorouridine
RTECSNumber YU8050000
TSCA No