CAS RN 316-41-6
CAS RN 316-41-6
IUPAC Name:
bis(16,17-dimethoxy-5,7-dioxa-13λ⁵-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁵,²⁰]henicosa-1(21),2,4(8),9,13,15,17,19-octaen-13-ylium) sulfate
Synonyms:
bis(berberine) sulfate
Molecular Weight (g/mol):
768.79
Molecular Formula:
C40H36N2O12S
InChi Key:
OJVABJMSSDUECT-UHFFFAOYSA-L
SMILES:
[O-]S([O-])(=O)=O.COC1=CC=C2C=C3C4=CC5=C(OCO5)C=C4CC[N+]3=CC2=C1OC.COC1=CC=C2C=C3C4=CC5=C(OCO5)C=C4CC[N+]3=CC2=C1OC
1
–
1
of
1
results
1
Berberine Sulfate Hydrate 98.0+%, TCI America™
CAS: 316-41-6 Molecular Formula: C40H44N2O15S+2 Molecular Weight (g/mol): 824.851 MDL Number: MFCD00031743 InChI Key: FYBWCLKVKGHJKS-UHFFFAOYSA-N PubChem CID: 129893573 SMILES: COC1=C(C2=C[N+]3=C(C=C2C=C1)C4=CC5=C(C=C4CC3)OCO5)OC.COC1=C(C2=C[N+]3=C(C=C2C=C1)C4=CC5=C(C=C4CC3)OCO5)OC.O.O.O.OS(=O)(=O)O