accessibility menu, dialog, popup

UserName

CAS RN 3147-55-5

IUPAC Name: 3,5-dibromo-2-hydroxybenzoic acid
Synonyms: 3,5-dibromo-2-hydroxybenzoic acid
Molecular Weight (g/mol): 295.91
Molecular Formula: C7H4Br2O3
InChi Key: BFBZHSOXKROMBG-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Br)=CC(Br)=C1O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)