missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromosalicylic Acid 98.0+%, TCI America™

Catalog No. D1087500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1087500G 500 g
D108725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1087500G Supplier TCI America Supplier No. D1087500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3147-55-5
Molecular Formula C7H4Br2O3
Molecular Weight (g/mol) 295.914
MDL Number MFCD00002441
InChI Key BFBZHSOXKROMBG-UHFFFAOYSA-N
Synonym 3,5-dibromosalicylic acid, benzoic acid, 3,5-dibromo-2-hydroxy, 2-hydroxy-3,5-dibromobenzoic acid, unii-t5jo5ua21h, salicylic acid, 3,5-dibromo, t5jo5ua21h, rarechem al bo 2002, attercop-chm at113669, labotest-bb lt00455167, labotest-bb lt03333482
PubChem CID 18464
IUPAC Name 3,5-dibromo-2-hydroxybenzoic acid
SMILES C1=C(C=C(C(=C1Br)O)C(=O)O)Br

Specifications

Melting Point 223°C
Color White-Yellow
Quantity 500 g
Formula Weight 295.91
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromosalicylic Acid