CAS RN 312700-07-5
IUPAC Name:
4-(9H-carbazol-9-yl)aniline hydrochloride
Synonyms:
4-(carbazol-9-yl)aniline hydrochloride
Molecular Weight (g/mol):
294.78
Molecular Formula:
C18H15ClN2
InChi Key:
CFQROJKMLJRSAL-UHFFFAOYSA-N
SMILES:
Cl.NC1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
Promotions Available
9-(4-Aminophenyl)carbazole hydrochloride, 98%
9-(4-Aminophenyl)carbazole hydrochloride, 98%
CAS: 312700-07-5 Molecular Formula: C18H15ClN2 Molecular Weight (g/mol): 294.782 MDL Number: MFCD00082678 InChI Key: CFQROJKMLJRSAL-UHFFFAOYSA-N Synonym: 4-9h-carbazol-9-yl aniline hydrochloride,9-4-aminophenyl carbazole hydrochloride,4-carbazol-9-yl aniline hydrochloride,4-carbazol-9-ylaniline hydrochloride,acmc-1agoy PubChem CID: 2794996 IUPAC Name: 4-carbazol-9-ylaniline;hydrochloride SMILES: C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=C(C=C4)N.Cl