accessibility menu, dialog, popup

UserName

CAS RN 3098-92-8

H2COO(E)

CAS RN 3098-92-8

IUPAC Name: ethenyl (2E)-3-phenylprop-2-enoate
Synonyms: ethenyl (2E)-3-phenylprop-2-enoate
Molecular Weight (g/mol): 174.2
Molecular Formula: C11H10O2
InChi Key: WGXGKXTZIQFQFO-CMDGGOBGSA-N
SMILES: C=COC(=O)\C=C\C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results