accessibility menu, dialog, popup

UserName

ClSOCl

CAS RN 3085-42-5

IUPAC Name: 1-chloro-4-(4-chlorobenzenesulfinyl)benzene
Synonyms: sulfoxide, bis(p-chlorophenyl)
Molecular Weight (g/mol): 271.16
Molecular Formula: C12H8Cl2OS
InChi Key: KJGYFISADIZFEL-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)S(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results