accessibility menu, dialog, popup

UserName

CAS RN 307531-83-5

IUPAC Name: zinc(2+) 2-(1,3-dioxolan-2-yl)ethanide bromide
Synonyms: zinc(2+) 2-(1,3-dioxolan-2-yl)ethanide bromide
Molecular Weight (g/mol): 246.41
Molecular Formula: C5H9BrO2Zn
InChi Key: FBIHMCAQTQOHLI-UHFFFAOYSA-M
SMILES: [Zn++].[Br-].[CH2-]CC1OCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results