accessibility menu, dialog, popup

UserName

CAS RN 30725-00-9

IUPAC Name: 6-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-one
Synonyms: 6-(hydroxymethyl)-2,2-dimethyl-dihydro-3aH-furo[3,4-d][1,3]dioxol-4-one
Molecular Weight (g/mol): 188.18
Molecular Formula: C8H12O5
InChi Key: NHHKFJCWLPPNCN-UHFFFAOYNA-N
SMILES: CC1(C)OC2C(CO)OC(=O)C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results